Methyl (2e)-3-cyclopentylprop-2-enoate 95%
Supplier: Apollo Scientific
OR958922-1G
OR958922-250MG
APOSOR958922-1GEA
571
GBP
APOSOR958922-1G
APOSOR958922-250MG
Methyl (2e)-3-cyclopentylprop-2-enoate 95%
Methyl (2e)-3-cyclopentylprop-2-enoate
Formula:
C₉H₁₄O₂ MW: 154,206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24134929 CAS Number: 136823-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...