4-Ethyl-3-nitroaniline 95%

Supplier: Apollo Scientific

Synonyms: 4-Ethyl-3-nitroaniline 4-Ethyl-3-nitroaniline

OR958761-1G OR958761-250MG
APOSOR958761-1GEA 70.9 GBP
APOSOR958761-1G APOSOR958761-250MG
4-Ethyl-3-nitroaniline 95%
4-Ethyl-3-nitroaniline
Formula: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971452
CAS Number: 51529-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR