(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride

Supplier: Apollo Scientific

Synonyms: Fmoc-Lys-OMe.HCl, Fmoc-Lys-OMe·HCl

OR958053-1G OR958053-250MG
APOSOR958053-1GEA 291 GBP
APOSOR958053-1G APOSOR958053-250MG
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride
Formula: C₂₂H₂₆N₂O₄·ClH
MW: 418,914 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275774
CAS Number: 847658-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR