Methyl 2-methyl-1,3-thiazole-5-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 2-methylthiazole-5-carboxylate

OR955745-1G OR955745-5G
APOSOR955745-1GEA 106 GBP
APOSOR955745-1G APOSOR955745-5G
Methyl 2-methyl-1,3-thiazole-5-carboxylate 95%
Methyl 2-methyl-1,3-thiazole-5-carboxylate
Formula: C₆H₇NO₂S
MW: 157,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14635883
CAS Number: 53233-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR