Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate 95%
Supplier: Apollo Scientific
|
Warning
|
OR955489-1G
OR955489-5G
APOSOR955489-1GEA
237
GBP
APOSOR955489-1G
APOSOR955489-5G
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate 95%
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate
Formula:
C₆H₁₁NO₃ MW: 145,156 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00797544 CAS Number: 114676-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...