3-Hydroxy-L-tyrosine methyl ester 96%

Supplier: Apollo Scientific
Warning

OR955473-1G OR955473-5G
APOSOR955473-1GEA 106 GBP
APOSOR955473-1G APOSOR955473-5G
3-Hydroxy-L-tyrosine methyl ester 96%
3-Hydroxy-L-tyrosine methyl ester
Formula: C₁₀H₁₃NO₄
MW: 211,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00870911
CAS Number: 7101-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR