3-Chloro-2-methyl-benzoic acid methyl ester 95%
Supplier: Apollo Scientific
Synonyms:
Methyl 3-chloro-2-methylbenzoate
OR955456-1G
OR955456-5G
APOSOR955456-1GEA
77.4
GBP
APOSOR955456-1G
APOSOR955456-5G
3-Chloro-2-methyl-benzoic acid methyl ester 95%
3-Chloro-2-methyl-benzoic acid methyl ester
Formula:
C₉H₉ClO₂ MW: 184,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00230148 CAS Number: 99586-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...