3-(3-Chloro-phenyl)-acrylic acid methyl ester 98%

Supplier: Apollo Scientific

OR955418-1G OR955418-5G
APOSOR955418-1GEA 243 GBP
APOSOR955418-1G APOSOR955418-5G
3-(3-Chloro-phenyl)-acrylic acid methyl ester 98%
3-(3-Chloro-phenyl)-acrylic acid methyl ester
Formula: C₁₀H₉ClO₂
MW: 196,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02149457
CAS Number: 42175-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR