2-Methyl-4-phenylphenol 95%

Supplier: Apollo Scientific

Synonyms: 3-Methyl-[1,1'-biphenyl]-4-ol

OR954881-1G OR954881-250MG
APOSOR954881-1GEA 551 GBP
APOSOR954881-1G APOSOR954881-250MG
2-Methyl-4-phenylphenol 95%
2-Methyl-4-phenylphenol
Formula: C₁₃H₁₂O
MW: 184,234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18313070
CAS Number: 30451-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR