4-Bromo-2-chloro-1-(methylsulphonyl)benzene 96%

Supplier: Apollo Scientific
Warning

OR954476-1G OR954476-5G
APOSOR954476-1GEA 154 GBP
APOSOR954476-1G APOSOR954476-5G
4-Bromo-2-chloro-1-(methylsulphonyl)benzene 96%
4-Bromo-2-chloro-1-(methylsulphonyl)benzene
Formula: C₇H₆BrClO₂S
MW: 269,543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26131172
CAS Number: 648905-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR