2-Chloroquinolin-6-ol 98%
Supplier: Apollo Scientific
OR954239-1G
OR954239-5G
APOSOR954239-1GEA
22
GBP
APOSOR954239-1G
APOSOR954239-5G
2-Chloroquinolin-6-ol 98%
2-Chloroquinolin-6-ol
Formula:
C₉H₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD12405093 CAS Number: 577967-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...