4-Chloro-2-methylpyrimidine-5-carboxylic acid 95%
Supplier: Apollo Scientific
OR954125-1G
OR954125-250MG
APOSOR954125-1GEA
584
GBP
APOSOR954125-1G
APOSOR954125-250MG
4-Chloro-2-methylpyrimidine-5-carboxylic acid 95%
4-Chloro-2-methylpyrimidine-5-carboxylic acid
Formula:
C₆H₅ClN₂O₂ MW: 172,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10000835 CAS Number: 933702-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...