Methyl 3-amino-5-methoxybenzoate 95%
Supplier: Apollo Scientific
OR953964-1G
OR953964-250MG
OR953964-5G
APOSOR953964-1GEA
90.5
GBP
APOSOR953964-1G
APOSOR953964-250MG
APOSOR953964-5G
Methyl 3-amino-5-methoxybenzoate 95%
Methyl 3-amino-5-methoxybenzoate
Formula:
C₉H₁₁NO₃ MW: 181,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09263221 CAS Number: 217314-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...