N-(Pyridin-3-ylmethyl)cyclopropanamine 95%

Supplier: Apollo Scientific

OR953961-1G OR953961-250MG
APOSOR953961-1GEA 328 GBP
APOSOR953961-1G APOSOR953961-250MG
N-(Pyridin-3-ylmethyl)cyclopropanamine 95%
N-(Pyridin-3-ylmethyl)cyclopropanamine
Formula: C₉H₁₂N₂
MW: 148,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03821869
CAS Number: 183609-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR