(4-Formyl-2-methoxy-phenoxy)-acetonitrile 95%

Supplier: Apollo Scientific

OR953940-10G OR953940-1G OR953940-5G
APOSOR953940-10GEA 905 GBP
APOSOR953940-10G APOSOR953940-1G APOSOR953940-5G
(4-Formyl-2-methoxy-phenoxy)-acetonitrile 95%
(4-Formyl-2-methoxy-phenoxy)-acetonitrile
Formula: C₁₀H₉NO₃
MW: 191,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01038240
CAS Number: 342592-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR