Imidazo[1,2-a]pyridin-2-yl-acetic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-(Imidazo[1,2-a]pyridin-2-yl)acetic acid

OR953789-1G OR953789-250MG
APOSOR953789-1GEA 774 GBP
APOSOR953789-1G APOSOR953789-250MG
Imidazo[1,2-a]pyridin-2-yl-acetic acid 95%
Imidazo[1,2-a]pyridin-2-yl-acetic acid
Formula: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03434391
CAS Number: 19741-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR