Ethyl 2-(2-chlorophenyl)acetate 95%

Supplier: Apollo Scientific

OR953222-100G OR953222-10G OR953222-25G
APOSOR953222-100GEA 64.3 GBP
APOSOR953222-100G APOSOR953222-10G APOSOR953222-25G
Ethyl 2-(2-chlorophenyl)acetate 95%
Ethyl 2-(2-chlorophenyl)acetate
Formula: C₁₀H₁₁ClO₂
MW: 198,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264019
CAS Number: 40061-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR