3-(4-Methylpiperazin-1-yl)benzaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(4-Methylpiperazin-1-yl)benzaldehyde
OR953111-1G
OR953111-5G
APOSOR953111-1GEA
159
GBP
APOSOR953111-1G
APOSOR953111-5G
3-(4-Methylpiperazin-1-yl)benzaldehyde
3-(4-Methylpiperazin-1-yl)benzaldehyde
Formula:
C₁₂H₁₆N₂O MW: 204,268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09030500 CAS Number: 628325-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...