N-Ethyl-4-methoxybenzenemethanamine 95%

Supplier: Apollo Scientific

Synonyms: N-(4-Methoxybenzyl)ethanamine

OR952824-1G OR952824-5G
APOSOR952824-1GEA 119 GBP
APOSOR952824-1G APOSOR952824-5G
N-Ethyl-4-methoxybenzenemethanamine 95%
N-Ethyl-4-methoxybenzenemethanamine
Formula: C₁₀H₁₅NO
MW: 165,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026704
CAS Number: 22993-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR