Acetylamino-cyclopentyl-acetic acid 95%

Supplier: Apollo Scientific

OR952789-1G OR952789-250MG
APOSOR952789-1GEA 263 GBP
APOSOR952789-1G APOSOR952789-250MG
Acetylamino-cyclopentyl-acetic acid 95%
Acetylamino-cyclopentyl-acetic acid
Formula: C₉H₁₅NO₃
MW: 185,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796733
CAS Number: 2521-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR