3-O-Tolyloxy-propylamine 95%

Supplier: Apollo Scientific

OR952780-1G OR952780-250MG
APOSOR952780-1GEA 328 GBP
APOSOR952780-1G APOSOR952780-250MG
3-O-Tolyloxy-propylamine 95%
3-O-Tolyloxy-propylamine
Formula: C₁₀H₁₅NO
MW: 165.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05053655
CAS Number: 50911-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR