5-Chloroimidazo[1,2-a]pyrazine 96%
Supplier: Apollo Scientific
OR951845-1G
OR951845-250MG
APOSOR951845-1GEA
408
GBP
APOSOR951845-1G
APOSOR951845-250MG
5-Chloroimidazo[1,2-a]pyrazine 96%
5-Chloroimidazo[1,2-a]pyrazine
Formula:
C₆H₄ClN₃ MW: 153,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193447 CAS Number: 63744-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...