Methyl (4-methyl-1,3-thiazol-2-yl)acetate 96%
Supplier: Apollo Scientific
OR951618-1G
OR951618-250MG
APOSOR951618-1GEA
780
GBP
APOSOR951618-1G
APOSOR951618-250MG
Methyl (4-methyl-1,3-thiazol-2-yl)acetate 96%
Methyl (4-methyl-1,3-thiazol-2-yl)acetate
Formula:
C₇H₉NO₂S MW: 171,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840444 CAS Number: 117840-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...