2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Methoxycarbonyl)cyclopropanecarboxylic acid
OR951034-1G
OR951034-250MG
APOSOR951034-1GEA
964
GBP
APOSOR951034-1G
APOSOR951034-250MG
2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid 95%
2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid
Formula:
C₆H₈O₄ MW: 144,125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16817526 CAS Number: 13279-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...