1-(3-Chloropropyl)piperidin-4-ol 95%
Supplier: Apollo Scientific
OR949939-1G
OR949939-250MG
APOSOR949939-1GEA
571
GBP
APOSOR949939-1G
APOSOR949939-250MG
1-(3-Chloropropyl)piperidin-4-ol 95%
1-(3-Chloropropyl)piperidin-4-ol
Formula:
C₈H₁₆ClNO MW: 177,672 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09745980 CAS Number: 145285-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...