2-Ethyl-3-methylbutan-1-ol 95%

Supplier: Apollo Scientific

OR949862-1G OR949862-250MG
APOSOR949862-1GEA 150 GBP
APOSOR949862-1G APOSOR949862-250MG
2-Ethyl-3-methylbutan-1-ol 95%
2-Ethyl-3-methylbutan-1-ol
Formula: C₇H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00089635
CAS Number: 32444-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR