8-Chlorochroman-4-one 95%

Supplier: Apollo Scientific

OR948297-1G OR948297-250MG
APOSOR948297-1GEA 103 GBP
APOSOR948297-1G APOSOR948297-250MG
8-Chlorochroman-4-one 95%
8-Chlorochroman-4-one
Formula: C₉H₇ClO₂
MW: 182,604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03695451
CAS Number: 49701-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR