5-Chloro-7-methyl-1H-indole-3-carbaldehyde 95%

Supplier: Apollo Scientific

OR948155-1G OR948155-250MG
APOSOR948155-1GEA 748 GBP
APOSOR948155-1G APOSOR948155-250MG
5-Chloro-7-methyl-1H-indole-3-carbaldehyde 95%
5-Chloro-7-methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12962649
CAS Number: 15936-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR