Methyl 3-(hydroxymethyl)-5-methoxybenzoate 95%

Supplier: Apollo Scientific
Warning

OR948040-1G OR948040-250MG
APOSOR948040-1GEA 126 GBP
APOSOR948040-1G APOSOR948040-250MG
Methyl 3-(hydroxymethyl)-5-methoxybenzoate 95%
Methyl 3-(hydroxymethyl)-5-methoxybenzoate
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD11044548
CAS Number: 367519-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR