(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 95%

Supplier: Apollo Scientific

Synonyms: H-D-3-Cl-Ala-OMe·HCl

OR947974-1G OR947974-5G
APOSOR947974-1GEA 44.6 GBP
APOSOR947974-1G APOSOR947974-5G
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 95%
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride
Formula: C₄H₈ClNO₂·ClH
MW: 174,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066119
CAS Number: 112346-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR