3-Bromo-2-methylpropyl acetate 95%

Supplier: Apollo Scientific

OR946908-1G OR946908-250MG
APOSOR946908-1GEA 681 GBP
APOSOR946908-1G APOSOR946908-250MG
3-Bromo-2-methylpropyl acetate 95%
3-Bromo-2-methylpropyl acetate
Formula: C₆H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16620805
CAS Number: 98485-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR