1-Methyl-3-methylidenecyclobutane-1-carboxylic acid
Supplier: Apollo Scientific
OR946186-1G
OR946186-250MG
OR946186-500MG
APOSOR946186-1GEA
1630
GBP
APOSOR946186-1G
APOSOR946186-250MG
APOSOR946186-500MG
1-Methyl-3-methylidenecyclobutane-1-carboxylic acid
1-Methyl-3-methylidenecyclobutane-1-carboxylic acid
Formula:
C₇H₁₀O₂ MW: 126,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19231380 CAS Number: 286442-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...