3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%
Supplier: Apollo Scientific
OR945262-1G
OR945262-250MG
APOSOR945262-1GEA
794
GBP
APOSOR945262-1G
APOSOR945262-250MG
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Formula:
C₉H₇NO₄ MW: 193,156 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11868820 CAS Number: 24132-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...