C-(8-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine hydrochloride 95%

Supplier: Apollo Scientific

OR944230-1G OR944230-250MG
APOSOR944230-1GEA 728 GBP
APOSOR944230-1G APOSOR944230-250MG
C-(8-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine hydrochloride 95%
C-(8-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine hydrochloride
Formula: C₈H₁₀N₄·ClH
MW: 198,653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362299
CAS Number: 1341040-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR