2,3-Dihydro-2-methyl-4(1H)-quinolinone 95%

Supplier: Apollo Scientific

OR944030-1G OR944030-250MG
APOSOR944030-1GEA 728 GBP
APOSOR944030-1G APOSOR944030-250MG
2,3-Dihydro-2-methyl-4(1H)-quinolinone 95%
2,3-Dihydro-2-methyl-4(1H)-quinolinone
Formula: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13179342
CAS Number: 30448-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR