3-(Propan-2-yloxy)benzonitrile 95%

Supplier: Apollo Scientific

OR943093-1G OR943093-250MG
APOSOR943093-1GEA 394 GBP
APOSOR943093-1G APOSOR943093-250MG
3-(Propan-2-yloxy)benzonitrile 95%
3-(Propan-2-yloxy)benzonitrile
Formula: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09933572
CAS Number: 363185-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR