2-Bromo-1,3-dichloro-5-methoxybenzene 98%

Supplier: Apollo Scientific
Warning

OR942147-1G OR942147-250MG
APOSOR942147-1GEA 187 GBP
APOSOR942147-1G APOSOR942147-250MG
2-Bromo-1,3-dichloro-5-methoxybenzene 98%
2-Bromo-1,3-dichloro-5-methoxybenzene
Formula: C₇H₅BrCl₂O
MW: 255,924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26743677
CAS Number: 174913-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR