t-Boc-N-Amido-PEG3-CH2CO2H 97%

Supplier: Apollo Scientific

Synonyms: 2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azahexadecan-16-oic acid

OR941103-1G OR941103-250MG
APOSOR941103-1GEA 460 GBP
APOSOR941103-1G APOSOR941103-250MG
t-Boc-N-Amido-PEG3-CH2CO2H 97%
t-Boc-N-Amido-PEG3-CH2CO2H
Formula: C₁₃H₂₅NO₇
MW: 307,34 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22376773
CAS Number: 462100-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR