tert-Butyl 3-amino-1H-pyrazole-1-carboxylate 95%
Supplier: Apollo Scientific
OR941096-1G
OR941096-250MG
APOSOR941096-1GEA
715
GBP
APOSOR941096-1G
APOSOR941096-250MG
tert-Butyl 3-amino-1H-pyrazole-1-carboxylate 95%
tert-Butyl 3-amino-1H-pyrazole-1-carboxylate
Formula:
C₈H₁₃N₃O₂ MW: 183,208 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13194910 CAS Number: 863504-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...