4-Bromo-2-methyl-2H-1,2,3-triazole 95%
Supplier: Apollo Scientific
OR939839-100MG
OR939839-1G
OR939839-250MG
OR939839-5G
OR939839-10G
APOSOR939839-100MGEA
0
GBP
APOSOR939839-100MG
APOSOR939839-1G
APOSOR939839-250MG
APOSOR939839-5G
APOSOR939839-10G
4-Bromo-2-methyl-2H-1,2,3-triazole 95%
4-Bromo-2-methyl-2H-1,2,3-triazole
Formula:
C₃H₄BrN₃ MW: 161,988 g/mol Boiling Pt: 161…162 °C Melting Pt: 22 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD23378485 CAS Number: 16681-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...