4-Chloro-2-(methylsulphonyl)pyrimidine 95%
Supplier: Apollo Scientific
OR939797-10G
OR939797-1G
OR939797-5G
APOSOR939797-10GEA
434
GBP
APOSOR939797-10G
APOSOR939797-1G
APOSOR939797-5G
4-Chloro-2-(methylsulphonyl)pyrimidine 95%
4-Chloro-2-(methylsulphonyl)pyrimidine
Formula:
C₅H₅ClN₂O₂S MW: 192,623 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09701372 CAS Number: 97229-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...