2-(Aminomethyl)benzoic acid 95%

Supplier: Apollo Scientific

OR939761-1G OR939761-500MG
APOSOR939761-1GEA 395 GBP
APOSOR939761-1G APOSOR939761-500MG
2-(Aminomethyl)benzoic acid 95%
2-(Aminomethyl)benzoic acid
Formula: C₈H₉NO₂
MW: 151,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06212682
CAS Number: 25672-97-3
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR