2',3'-Dimethylacetophenone 95%
Supplier: Apollo Scientific
Synonyms:
1-(2,3-Dimethylphenyl)ethanone
OR939623-10G
OR939623-1G
OR939623-5G
APOSOR939623-10GEA
146
GBP
APOSOR939623-10G
APOSOR939623-1G
APOSOR939623-5G
2',3'-Dimethylacetophenone 95%
2',3'-Dimethylacetophenone
Formula:
C₁₀H₁₂O MW: 148,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03844735 CAS Number: 2142-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...