Ethylhydrazine hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR939620-1G OR939620-5G
APOSOR939620-1GEA 86.4 GBP
APOSOR939620-1G APOSOR939620-5G
Ethylhydrazine hydrochloride 95%
Formula: C₂H₈N₂·ClH
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD01722460
CAS Number: 18413-14-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR