3-Methyl-3-phenylbutanal 95%

Supplier: Apollo Scientific

OR939467-1G OR939467-250MG
APOSOR939467-1GEA 479 GBP
APOSOR939467-1G APOSOR939467-250MG
3-Methyl-3-phenylbutanal 95%
3-Methyl-3-phenylbutanal
Formula: C₁₁H₁₄O
MW: 162,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00156879
CAS Number: 6325-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR