1-Iodo-2-methoxy-ethane 97%

Supplier: Apollo Scientific

Synonyms: 1-Iodo-2-methoxyethane

OR938738-1G OR938738-5G
APOSOR938738-1GEA 307 GBP
APOSOR938738-1G APOSOR938738-5G
1-Iodo-2-methoxy-ethane 97%
1-Iodo-2-methoxy-ethane
Formula: C₃H₇IO
MW: 185,992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07644549
CAS Number: 4296-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR