Sodium methanethiosulphonate 95%
Supplier: Apollo Scientific
OR938724-1G
OR938724-5G
APOSOR938724-1GEA
0
GBP
APOSOR938724-1G
APOSOR938724-5G
Sodium methanethiosulphonate 95%
Sodium methanethiosulphonate
Formula:
CH₃NaO₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD03425630 CAS Number: 1950-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...