1-(Aminomethyl)-cycloheptanol 97%

Supplier: Apollo Scientific

OR938662-1G OR938662-250MG
APOSOR938662-1GEA 895 GBP
APOSOR938662-1G APOSOR938662-250MG
1-(Aminomethyl)-cycloheptanol 97%
1-(Aminomethyl)-cycloheptanol
Formula: C₈H₁₇NO
MW: 143.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835359
CAS Number: 45732-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR