5-Isopropyl-2H-pyrazole-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR938480-1G
OR938480-5G
APOSOR938480-1GEA
55.1
GBP
APOSOR938480-1G
APOSOR938480-5G
5-Isopropyl-2H-pyrazole-3-carboxylic acid 95%
5-Isopropyl-2H-pyrazole-3-carboxylic acid
Formula:
C₇H₁₀N₂O₂ MW: 154,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02224142 CAS Number: 92933-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...