2-(Cyclopentyloxy)nicotinonitrile 97%

Supplier: Apollo Scientific

OR937961-1G OR937961-5G
APOSOR937961-1GEA 197 GBP
APOSOR937961-1G APOSOR937961-5G
2-(Cyclopentyloxy)nicotinonitrile 97%
2-(Cyclopentyloxy)nicotinonitrile
Formula: C₁₁H₁₂N₂O
MW: 188,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09946128
CAS Number: 1016805-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR